missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ (-)-Quinpirole hydrochloride

Description
Selective dopamine D2 receptor agonist (Ki values are 4.8, ∽24, ∽30 and 1900 nM at D2, D3, D4 and D1 receptors respectively).
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 255.79 |
| Purity | >98% |
| Molecular Weight (g/mol) | 255.79 |
| Chemical Name or Material | (-)-Quinpirole hydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?