missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-(2-Bromophenyl)ethanol, 98%

Product Code. p-7037295
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15452418

Brand: Thermo Scientific Alfa Aesar H56003.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 76116-20-6 | C8H9BrO | 201.063 g/mol

It is the most important dyestuff intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is the most important dyestuff intermediate.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76116-20-6
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.063
MDL Number MFCD00216591
InChI Key DZLZSFZSPIUINR-ZCFIWIBFSA-N
Synonym r-1-2-bromophenyl ethanol, 1r-1-2-bromophenyl ethanol, 1r-1-2-bromophenyl ethan-1-ol, pubchem5686, r-+-1-2-bromophenyl ethanol, r-+-2-bromo-, a-methylbenzyl alcohol, benzenemethanol,2-bromo-a-methyl-, ar, r-+-2-bromo-alpha-methylbenzyl alcohol, r-+-2-bromo-alpha-methylbenzyl alcoh
PubChem CID 2734868
IUPAC Name (1R)-1-(2-bromophenyl)ethanol
SMILES CC(C1=CC=CC=C1Br)O

Specifications

Melting Point 54°C to 56°C
Quantity 250 mg
Optical Rotation +54° (c=1 in Chloroform)
Formula Weight 201.06
Percent Purity 98%
Chemical Name or Material (R)-(+)-1-(2-Bromophenyl)ethanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.