missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-[(4-Chlorophenyl)phenylmethyl]piperazine, TRC

Product Code. 30136282
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136282 25 mg 25mg
30135245 50 mg 50mg
30007148 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136282 Supplier Toronto Research Chemicals Supplier No. TRCC37957425MG
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 300543-56-0
Molecular Formula C17 H19 Cl N2
Molecular Weight (g/mol) 286.799
Synonym Piperazine, 1-[(R)-(4-chlorophenyl)phenylmethyl]-, (-)-1-[(4-Chlorophenyl)phenylmethyl]piperazine, (R)-1-(p-Chlorobenzhydryl)piperazine, 1-[(R)-α-(4-Chlorophenylbenzyl]piperazine, N-[(R)-α-(4-Chlorophenylbenzyl]piperazine
IUPAC Name 1-[(R)-(4-chlorophenyl)-phenylmethyl]piperazine
SMILES Clc1ccc(cc1)[C@H](N2CCNCC2)c3ccccc3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H19ClN2/c18-16-8-6-15(7-9-16)17(14-4-2-1-3-5-14)20-12-10-19-11-13-20/h1-9,17,19H,10-13H2/t17-/m1/s1
Formula Weight 286.124
Percent Purity >95%
Chemical Name or Material (R)-1-[(4-Chlorophenyl)phenylmethyl]piperazine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.