missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-(4-Methoxyphenyl)ethylamine, ChiPros 99+%, ee 99+%, Thermo Scientific™

Product Code. 11382997
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11382997 25 g 25g
11362997 1 g 1g
11372997 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 11382997

Brand: Thermo Scientific Alfa Aesar L16321.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(R)-(+)-1-(4-Methoxyphenyl)ethylamine use in diastereo-and enantioselective Michael addition reactions. Thiazoles has been prepared from (R)-(+)-1-(4-Methoxyphenyl)ethylamine.

Solubility
Soluble in water (10g/L).

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22038-86-4
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00671659
InChI Key JTDGKQNNPKXKII-SSDOTTSWSA-N
Synonym r-+-1-4-methoxyphenyl ethylamine, r-1-4-methoxyphenyl ethylamine, r-1-4-methoxyphenyl ethanamine, 1r-1-4-methoxyphenyl ethanamine, r-+-4-methoxy-alpha-methylbenzylamine, s-4-methoxy-alpha-methylbenzylamine, r +-1-4-methylphenyl ethylamine, pubchem9920, ksc496q8b, rm+-1-4-methoxyphenyl ethylamine
PubChem CID 641449
IUPAC Name (1R)-1-(4-methoxyphenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)OC)N

Specifications

Melting Point <-20°C
Density 1.02
pH 11
Boiling Point 65°C (0.38 mmHg)
Odor Amine-like
Refractive Index 1.533
Quantity 25 g
UN Number UN2735
Beilstein 2413029
Sensitivity Air Sensitive
Solubility Information Soluble in water (10g/L).
Optical Rotation +34° (Neat)
Formula Weight 151.21
Percent Purity ≥99%
Assay ee ≥99%
Chemical Name or Material (R)-(+)-1-(4-Methoxyphenyl)ethylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.