missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-[alpha-(4-Chlorophenyl)benzyl]piperazine, 97%, Thermo Scientific Chemicals

Product Code. 15460559
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15460559 5 g 5g
15478476 250 mg 250mg
15450559 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15460559 Supplier Thermo Scientific Alfa Aesar Supplier No. H63722.06
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

(R)-1-[alpha-(4-Chlorophenyl)benzyl]piperazine intermediate in the production of (R)-Cetirizine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-1-[alpha-(4-Chlorophenyl)benzyl]piperazine intermediate in the production of (R)-Cetirizine.

Solubility
Soluble in chloroform, methanol, and dichloromethane. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 300543-56-0
Molecular Formula C17H19ClN2
Molecular Weight (g/mol) 286.803
MDL Number MFCD11519277
InChI Key UZKBSZSTDQSMDR-QGZVFWFLSA-N
Synonym r-1-4-chlorophenyl phenyl methyl piperazine, r---1-4-chlorophenyl phenylmethyl piperazine, norchlorcyclizine,-, unii-890a9g29pg, r-1-4-chlorophenyl phenylmethyl piperazine, r-1-p-chlorobenzhydryl piperazine, 1-r-4-chlorophenyl phenyl methyl piperazine, piperazine, 1-r-4-chlorophenyl phenylmethyl, 1-r-4-chlorophenyl-phenylmethyl piperazine
PubChem CID 668697
IUPAC Name 1-[(R)-(4-chlorophenyl)-phenylmethyl]piperazine
SMILES C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl

Specifications

Color White
Quantity 5 g
Solubility Information Soluble in chloroform,methanol,and dichloromethane. Slightly soluble in water.
Formula Weight 286.8
Percent Purity 97%
Physical Form Solid
Chemical Name or Material (R)-1-[alpha-(4-Chlorophenyl)benzyl]piperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.