missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-Phenylpropylamine, ChiPros 99+%, ee 98%

Product Code. 11322997
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322997 25 g 25g
11302997 1 g 1g
11312997 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11322997 Supplier Thermo Scientific Alfa Aesar Supplier No. L16319.14
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3082-64-2 | C9H13N | 135.21 g/mol

For synthesis of optically active products

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3082-64-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00083057
InChI Key AQFLVLHRZFLDDV-SECBINFHSA-N
Synonym r-+-1-phenylpropylamine, r-1-phenylpropan-1-amine, 1r-1-phenylpropan-1-amine, r-1-phenylpropylamine, r-+-alpha-ethylbenzylamine, 1r-1-phenylpropylamine, r-+-methylbenzylamine, r-+-1-amino-1-phenylpropane, r +-a-ethylbenzylamine, r-+-, a-ethylbenzylamine
PubChem CID 5324978
IUPAC Name (1R)-1-phenylpropan-1-amine
SMILES CCC(C1=CC=CC=C1)N

Specifications

Melting Point -69°C
Density 0.93
Boiling Point 205°C
Flash Point 77°C (170°F)
Odor Amine-like
Refractive Index 1.52
Quantity 25 g
UN Number UN2735
Beilstein 3029904
Sensitivity Air Sensitive
Optical Rotation +20° (Neat)
Formula Weight 135.21
Percent Purity ≥99%
Assay ee 98%
Chemical Name or Material (R)-(+)-1-Phenylpropylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.