missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Phenylpropionic acid, 99%

Product Code. 10430141 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass Bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10430141 250 mg Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10430141 Supplier Thermo Scientific Acros Supplier No. 264662500
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7782-26-5 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7782-26-5
CAS Min % 98.5
CAS Max % 100.0
Melting Point 29°C to 32°C
Color Beige to Brown
Density 1.1000g/mL
Boiling Point 115°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C9H10O2
Linear Formula CH3CH(C6H5)CO2H
Refractive Index 1.5232
MDL Number MFCD00063140
Quantity 250 mg
Beilstein 09, 2417
Specific Gravity 1.1
Specific Rotation Condition −75° (20°C c=3,CHCl3)
Specific Rotation -75
Synonym r---2-phenylpropionic acid, r-2-phenylpropanoic acid, 2r-2-phenylpropanoic acid, r-hydratropic acid, --hydratropic acid, r---hydratropic acid, r---2-phenylpropionicacid, r-2-phenylpropionic acid, r-2-phenyl-propionic acid, r-2-phenyl-proprionic acid
InChI Key YPGCWEMNNLXISK-SSDOTTSWSA-N
SMILES CC(C1=CC=CC=C1)C(=O)O
IUPAC Name (2R)-2-phenylpropanoic acid
Molecular Weight (g/mol) 150.18
PubChem CID 446626
ChEBI CHEBI:43035
Formula Weight 150.18
Percent Purity 99%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material (R)-(-)-2-Phenylpropionic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(-)-2-Phenylpropionic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.