missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-3-Aminotetrahydrofuran p-toluenesulfonate salt, 95%, Thermo Scientific Chemicals

Product Code. 10159699 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10159699 5 g 5g
10453520 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10159699 Supplier Thermo Scientific Acros Supplier No. 435820050
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 111769-27-8 | Molecular Formula: C11H17NO4S | Molecular Weight: 259.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111769-27-8
Molecular Formula C11H17NO4S
Molecular Weight (g/mol) 259.32
MDL Number MFCD00040622,MFCD00040622,MFCD00066238
InChI Key BZXPLADBSZWDIH-UHFFFAOYNA-N
Synonym r-3-aminotetrahydrofuran tosylate, r-+-tetrahydro-3-furylamine p-toluenesulfonate salt, r---3-aminotetrahydrofuran toluene-4-sulfonate, r-+-3-aminotetrahydrofuran toluene-4-sulfonate, r---3-aminotetrahydrofurantoluene-4-sulfonate, 3-furanamine, tetrahydro-, r-, 4-methylbenzenesulfonate, c4h9no.c7h8o3s, r-tetrahydrofuran-3-amine-4-methylbenzenesulfonate, 3r-oxolan-3-amine; para-toluene sulfonate, 4-methylbenzenesulfonic acid; 3r-tetrahydrofuran-3-amine
PubChem CID 14243169
IUPAC Name 4-methylbenzenesulfonic acid;(3R)-oxolan-3-amine
SMILES NC1CCOC1.CC1=CC=C(C=C1)S(O)(=O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 137.3°C to 141°C
Color White
Elemental Analysis 48.91 to 52.99% C, 5.18 to 5.62% N
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 5 g
Specific Rotation 4.3
Formula Weight 259.32
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-3-Aminotetrahydrofuran p-toluenesulfonate salt
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(+)-3-Aminotetrahydrofuran p-toluenesulfonate salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.