missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(Boc-amino)-3-(3-chlorophenyl)propionic acid, 98%, Thermo Scientific™

Product Code. 15459495
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459495 250 mg 250mg
15439495 1 g 1g
15449495 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15459495 Supplier Thermo Scientific Alfa Aesar Supplier No. H52003.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500789-06-0
Molecular Formula C14H18ClNO4
Molecular Weight (g/mol) 299.751
MDL Number MFCD03427959
InChI Key UDUKZORPLJUWTF-LLVKDONJSA-N
Synonym r-3-tert-butoxycarbonyl amino-3-3-chlorophenyl propanoic acid, boc-r-3-amino-3-3-chlorophenyl propionic acid, r-3-boc-amino-3-3-chlorophenyl propionic acid, 3r-3-tert-butoxy carbonylamino-3-3-chlorophenyl propanoic acid, 3r-3-tert-butoxycarbonyl amino-3-3-chlorophenyl propanoic acid, boc-d-?-phe 3-cl-oh, boc-d-beta-phe 3-cl-oh, boc-phg 3-cl-c*ch2 oh, r-boc-3-chloro-beta-phenylalanine, boc-d-3-amino-3-3-chlorophenyl-propionic acid
PubChem CID 2761720
IUPAC Name (3R)-3-(3-chlorophenyl)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC(=CC=C1)Cl

Specifications

Melting Point 90°C to 92°C
Quantity 250 mg
Optical Rotation +40° (c=1 in Ethanol)
Formula Weight 299.75
Percent Purity 98%
Chemical Name or Material (R)-3-(Boc-amino)-3-(3-chlorophenyl)propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.