missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Bromo-alpha-methylbenzyl alcohol, 95%, 98% ee

Product Code. 10730494 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10730494 5 g Glass bottle 5g
10526553 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10730494 Supplier Thermo Scientific Acros Supplier No. 376490050
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 134615-24-0 | C8H9BrO | 201.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134615-24-0
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD06658929
InChI Key ULMJQMDYAOJNCC-ZCFIWIBFSA-N
Synonym r-1-3-bromophenyl ethanol, 1r-1-3-bromophenyl ethanol, 1r-1-3-bromophenyl ethan-1-ol, pubchem15565, r-1-m-bromophenyl ethanol, r-1-3-bromo-phenyl-ethanol, r-3-bromo-alpha-methylbenzyl alcohol, benzenemethanol, 3-bromo-a-methyl-, ar
PubChem CID 2734889
IUPAC Name (1R)-1-(3-bromophenyl)ethanol
SMILES CC(C1=CC(=CC=C1)Br)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OH)C6H4Br
Quantity 5 g
Formula Weight 201.06
Percent Purity 95%,98% ee
Physical Form Liquid
Chemical Name or Material (R)-3-Bromo-α-methylbenzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • (R)-3-Bromo-a-methylbenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.