missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid, 98%

Product Code. 15499525
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499525 5 g 5g
15409535 250 mg 250mg
15489525 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15499525 Supplier Thermo Scientific Alfa Aesar Supplier No. H52037.06
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 517905-93-0 | C24H20N2O6 | 432.43 g/mol

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 517905-93-0
Molecular Formula C24H20N2O6
Molecular Weight (g/mol) 432.43
MDL Number MFCD03428045
InChI Key DRESTDPDCGPKNI-UHFFFAOYNA-N
Synonym r-3-9h-fluoren-9-yl methoxy carbonyl amino-3-2-nitrophenyl propanoic acid, fmoc-r-3-amino-3-2-nitrophenyl-propionic acid, fmoc-d-beta-phe 2-no2-oh, 3r-3-9h-fluoren-9-ylmethoxy carbonyl amino-3-2-nitrophenyl propanoic acid, pubchem24026, fmoc-beta-phe 2-no2-oh, fmoc-d-?-phe 2-no2-oh, n-fmoc-2-nitro-l-beta-phenylalanine, fmoc-r-3-amino-3-2-nitro-phenyl-propionic acid, r-3-fmoc-amino-3-2-nitrophenyl propionic acid
PubChem CID 2761743
IUPAC Name 3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(2-nitrophenyl)propanoic acid
SMILES OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 170°C to 172°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Optical Rotation −182° (c=1 in DMF)
Formula Weight 432.43
Percent Purity 98%
Chemical Name or Material (R)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.