missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, 99%

Product Code. 10190212 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10190212 250 mg Glass bottle 250mg
10555302 1 g Glass bottle 1g
10444215 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10190212 Supplier Thermo Scientific Acros Supplier No. 312222500
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 20445-31-2 | C10H9F3O3 | 234.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20445-31-2
Molecular Formula C10H9F3O3
Molecular Weight (g/mol) 234.17
MDL Number MFCD00004184
InChI Key JJYKJUXBWFATTE-UHFFFAOYNA-N
Synonym +-mtpa, unii-27o5l9t1wm, r-moshers acid, +-mosher's acid, 2r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, r-+-alpha-methoxy-alpha-trifluoromethylphenylacetic acid, r-mtpa, r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, r-+-a-methoxy-a-trifluoromethylphenylacetic acid, r-3,3,3-trifluoro-2-methoxy-2-phenyl-propionic acid
PubChem CID 2723917
IUPAC Name (2R)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid
SMILES COC(C(O)=O)(C1=CC=CC=C1)C(F)(F)F

Specifications

Melting Point 46.0°C to 49.0°C
Color White
Boiling Point 116.0°C to 118.0°C (1.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C6H5C(OCH3)(CF3)CO2H
Quantity 250 mg
Fieser 11,329
Merck Index 15,6365
Specific Rotation + 69.00
Formula Weight 234.17
Percent Purity 99%(e.e.>99.5%)
Physical Form Crystalline Low Melting Solid
Chemical Name or Material (R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid, 99%(e.e.>99.5%)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.