missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, 99%

Product Code. 11406126
Change view
Click to view available options
Quantity:
0.1 g
500 mg
2.5 g
Unit Size:
0.1g
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11406126 2.5 g 2.5g
11443990 500 mg 500mg
11416126 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11406126 Supplier Thermo Scientific Alfa Aesar Supplier No. B22968.88
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 20445-31-2 | C10H9F3O3 | 234.17 g/mol

(R)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid is widely used reagent for determination of optical purities of alcohols and amines by 1H or 19F NMR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid is widely used reagent for determination of optical purities of alcohols and amines by 1H or 19F NMR.

Solubility
Soluble in methanol (50 mg/ 1 ml).

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20445-31-2
Molecular Formula C10H9F3O3
Molecular Weight (g/mol) 234.17
MDL Number MFCD00004184
InChI Key JJYKJUXBWFATTE-UHFFFAOYNA-N
PubChem CID 2723917
SMILES COC(C(O)=O)(C1=CC=CC=C1)C(F)(F)F

Specifications

Melting Point 43°C to 45°C
Density 1.344
Boiling Point 105°C to 107°C (1 mmHg)
Flash Point >110°C
Refractive Index 1.473
Quantity 2.5 g
Beilstein 2215440
Sensitivity Hygroscopic
Merck Index 14,6280
Synonym +-mtpa, unii-27o5l9t1wm, r-moshers acid, +-mosher's acid, 2 r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, r-+-alpha-methoxy-alpha-trifluoromethylphenylacetic acid, r-mtpa, r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, r-+-a-methoxy-a-trifluoromethylphenylacetic acid, r-3,3,3-trifluoro-2-methoxy-2-phenyl-propionic acid
Solubility Information Soluble in methanol (50 mg/ 1 mL).
Optical Rotation +72°C=2 in. Methanol)
IUPAC Name (2 R)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid
Formula Weight 234.18
Percent Purity 99%
Chemical Name or Material (R)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.