missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 98+%

Product Code. 11353466
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353466 1 g 1g
11363466 50 mg 50mg
11373466 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11353466 Supplier Thermo Scientific Alfa Aesar Supplier No. L14325.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 39637-99-5 | C10H8ClF3O2 | 252.62 g/mol

(R)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride is used as a chiral acylating reagent, which is used in the resolution of amino acid enantiomers like 2,5-dimethoxy-4-methylamphetamine. It is also suitable for the determination of the optical composition of compounds extracted from biological fluids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride is used as a chiral acylating reagent, which is used in the resolution of amino acid enantiomers like 2,5-dimethoxy-4-methylamphetamine. It is also suitable for the determination of the optical composition of compounds extracted from biological fluids.

Solubility
Sparingly miscible with water.

Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39637-99-5
Molecular Formula C10H8ClF3O2
Molecular Weight (g/mol) 252.62
MDL Number MFCD00044400
InChI Key PAORVUMOXXAMPL-VIFPVBQESA-N
Synonym unii-81ut10uhv3, r---mtpa-cl, --alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, r---alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, 2r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, r---a-methoxy-a-trifluoromethylphenylacetyl chloride, r---alpha-methoxy-alpha-trifluoromethylphenylacetyl chloride, --mtpa-cl, --mosher's acid chloride
PubChem CID 3080792
IUPAC Name (2R)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride
SMILES CO[C@](C(Cl)=O)(C1=CC=CC=C1)C(F)(F)F

Specifications

Density 1.353
Boiling Point 213°C to 214°C
Flash Point 89°C (192°F)
Refractive Index 1.469
Quantity 1 g
UN Number UN3265
Beilstein 3591565
Sensitivity Moisture sensitive
Merck Index 14,6280
Solubility Information Sparingly miscible with water.
Optical Rotation −137° (Neat)
Formula Weight 252.62
Percent Purity ≥98%
Chemical Name or Material (R)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.