missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 98+%

Product Code. 11363466
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
This item is not returnable. View return policy

Product Code. 11363466

Brand: Thermo Scientific Alfa Aesar L14325.MF

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39637-99-5 | C10H8ClF3O2 | 252.62 g/mol

(R)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride is used as a chiral acylating reagent, which is used in the resolution of amino acid enantiomers like 2,5-dimethoxy-4-methylamphetamine. It is also suitable for the determination of the optical composition of compounds extracted from biological fluids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride is used as a chiral acylating reagent, which is used in the resolution of amino acid enantiomers like 2,5-dimethoxy-4-methylamphetamine. It is also suitable for the determination of the optical composition of compounds extracted from biological fluids.

Solubility
Sparingly miscible with water.

Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39637-99-5
Molecular Formula C10H8ClF3O2
Molecular Weight (g/mol) 252.62
MDL Number MFCD00044400
InChI Key PAORVUMOXXAMPL-VIFPVBQESA-N
Synonym unii-81ut10uhv3, r---mtpa-cl, --alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, r---alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, 2r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, r---a-methoxy-a-trifluoromethylphenylacetyl chloride, r---alpha-methoxy-alpha-trifluoromethylphenylacetyl chloride, --mtpa-cl, --mosher's acid chloride
PubChem CID 3080792
IUPAC Name (2R)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride
SMILES CO[C@](C(Cl)=O)(C1=CC=CC=C1)C(F)(F)F

Specifications

Density 1.353
Boiling Point 213°C to 214°C
Flash Point 89°C (192°F)
Refractive Index 1.469
Quantity 50 mg
UN Number UN3265
Beilstein 3591565
Sensitivity Moisture sensitive
Merck Index 14,6280
Solubility Information Sparingly miscible with water.
Optical Rotation −137° (Neat)
Formula Weight 252.62
Percent Purity ≥98%
Chemical Name or Material (R)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.