missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-alpha-Methoxy-alpha-trifluoromethylphenylacetyl chloride, Maybridge

Product Code. 10188224
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10188224 1 g 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10188224 Supplier Thermo Scientific Maybridge Supplier No. MAY00230DA
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 39637-99-5
Density 1.3400g/mL
Assay Percent Range 97+%
Molecular Formula C10H8ClF3O2
MDL Number MFCD00044400
Quantity 1 g
Merck Index 15, 6365
Specific Rotation Condition −132.00 (20.00°C c=6,CHCl3)
Synonym unii-81ut10uhv3, r---mtpa-cl, --alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, r---alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, 2r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, r---a-methoxy-a-trifluoromethylphenylacetyl chloride, r---alpha-methoxy-alpha-trifluoromethylphenylacetyl chloride, --mtpa-cl, --mosher's acid chloride
InChI Key PAORVUMOXXAMPL-VIFPVBQESA-N
SMILES CO[C@](C(Cl)=O)(C1=CC=CC=C1)C(F)(F)F
IUPAC Name (2R)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride
Molecular Weight (g/mol) 252.62
PubChem CID 3080792
Formula Weight 252.62
Percent Purity 97%
Chemical Name or Material (R)-(-)-α-Methoxy-α-trifluoromethylphenylacetyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.