missing translation for 'onlineSavingsMsg'
Learn More

(R)-Cetirizine Dihydrochloride, TRC

Product Code. 30003490
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003490 25 mg 25mg
30008846 250 mg 250mg
30009148 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003490 Supplier Toronto Research Chemicals Supplier No. TRCC28110625MG
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130018-87-0
Molecular Formula C21 H25 Cl N2 O3 . 2 Cl H
Molecular Weight (g/mol) 461.81
Synonym Levocetirizine Dihydrochloride, Acetic acid, 2-[2-[4-[(R)-(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-, hydrochloride (1:2), Acetic acid, [2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-, dihydrochloride, (R)-, Acetic acid, [2-[4-[(R)-(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-, dihydrochloride (9CI), (R)-(-)-Cetirizine hydrochloride, (R)-Cetirizine dihydrochloride, Allear, Allercet, Levocetirizine dihydrochloride, Lezyncet, UCB 28556, Xusal, Xyzal, Xyzall
IUPAC Name 2-[2-[4-[(R)-(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetic acid;dihydrochloride
SMILES Cl.Cl.OC(=O)COCCN1CCN(CC1)[C@H](c2ccccc2)c3ccc(Cl)cc3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C21H25ClN2O3.2ClH/c22-19-8-6-18(7-9-19)21(17-4-2-1-3-5-17)24-12-10-23(11-13-24)14-15-27-16-20(25)26;;/h1-9,21H,10-16H2,(H,25,26);2*1H/t21-;;/m1../s1
Formula Weight 460.1087
Percent Purity >95%
Chemical Name or Material Levocetirizine Dihydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.