missing translation for 'onlineSavingsMsg'
Learn More

(R)-Clodinafop, TRC

Product Code. 30360434
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30360434 10 mg
30355828 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30360434 Supplier Toronto Research Chemicals Supplier No. TRCC58450010MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114420-56-3
Molecular Formula C14 H11 Cl F N O4
Molecular Weight (g/mol) 311.69
Synonym Propanoic acid, 2-[4-[(5-chloro-3-fluoro-2-pyridinyl)oxy]phenoxy]-, (R)-, (2R)-2-[4-[(5-Chloro-3-fluoro-2-pyridinyl)oxy]phenoxy]propanoic acid, CGA 193469, Clodinafop
IUPAC Name (2R)-2-[4-(5-chloro-3-fluoropyridin-2-yl)oxyphenoxy]propanoic acid
SMILES C[C@@H](Oc1ccc(Oc2ncc(Cl)cc2F)cc1)C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H11ClFNO4/c1-8(14(18)19)20-10-2-4-11(5-3-10)21-13-12(16)6-9(15)7-17-13/h2-8H,1H3,(H,18,19)/t8-/m1/s1
Formula Weight 311.0361
Chemical Name or Material Clodinafop free acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.