missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Cultrex Basement Membrane Extract, PathClear
Click to view available options
Quantity:
1 mL
5 mL
2 x 5 mL
Conditionnement:
10mL
1mL
5mL
Description
Functional Assay: Tube formation - BME promotes formation of capillary-like structures by human (HBMVEC; HUVEC) or mouse (SVEC4-10) endothelial cells. Gelling Assay: BME gels in less than 30 minutes at 37degreesC, and maintains the gelled form in culture medium for a minimum of 14 days at 37degreesC.
Spécification
Spécification
| Concentration | 12 to 18 mg/mL |
| For Use With (Application) | Bioactivity |
| Quantity | 2 x 5 mL |
| Endotoxin Level | ≤8EU/mL |
| Source | Murine Engelbreth-Holm-Swarm (EHS) tumor. |
| Content And Storage | Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu