missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Proteome Purify 2 Mouse Serum Protein Immunodepletion Resin

Product Code. 18038101
Change view
Click to view available options
Quantity:
20 Tests
40 Tests
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18038101 40 Tests 1 set
18078181 20 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18038101 Supplier R&D Systems™ Supplier No. MIDR002040
  • ONLINE EXCLUSIVE PRICE
    1205.00 €
    1137.15 € / 1 sets
    Save 67.85 €
    5% Off
 Restricted Item
Estimated Shipment: 15-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Provides an easy-to-use, high capacity system to reproducibly remove albumin and IgG from mouse serum or plasma

Analysis of low abundance proteins by even the most sensitive of methods for proteomics greatly benefits from the removal of high abundance proteins that may interfere with their detection.

  • Improves visualization of target proteins
  • Allows user to increase protein load or concentration

TRUSTED_SUSTAINABILITY

Specifications

Quantity 40 Tests
Product Type Immunodepletion Resin
Content And Storage Store the unopened product at 2 - 8° C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.