missing translation for 'onlineSavingsMsg'
Learn More

(R)-Epichlorohydrin, TRC

Product Code. 30382335
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382335 5 g -
30382427 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30382335 Supplier Toronto Research Chemicals Supplier No. TRCE5823055G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51594-55-9
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.52
Synonym (2R)-(Chloromethyl)oxirane, (-)-2-(Chloromethyl)oxirane, (-)-Epichlorohydrin, (2R)-2-(Chloromethyl)oxirane, (R)-(-)-Epichlorohydrin, (R)-(-)-Epichlorohydrin, (R)-(Chloromethyl)oxirane, (R)-1-Chloro-2,3-epoxypropane
IUPAC Name (2R)-2-(chloromethyl)oxirane
SMILES ClC[C@H]1CO1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C3H5ClO/c4-1-3-2-5-3/h3H,1-2H2/t3-/m0/s1
Formula Weight 92.0029
Chemical Name or Material (R)-Epichlorohydrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.