missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R-IMPP, MedChemExpress
Description
R-IMPP (PF-00932239) is an anti-secretagogue of PCSK9 (IC50=4.8 μM), which targets the 80S ribosome to inhibit PCSK9 protein translation.
Specifications
Specifications
| Chemical Name or Material | R-IMPP |
| CAS | 2133832-83-2 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C24H27N3O2 |
| Quantity | 50 mg |
| Synonym | PF-00932239 |
| Solubility Information | DMSO : 100 mg/mL (256.75 mM; Need ultrasonic) |
| SMILES | O=C(N(C1=NC=CC2=C1C=CC=C2)[C@H]3CNCCC3)CCC4=CC=C(OC)C=C4 |
| Molecular Weight (g/mol) | 389.49 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?