missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Ketoprofen, TRC

Product Code. 30010257
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010257 10 mg 10mg
18042740 25 mg 25mg
18021130 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010257 Supplier Toronto Research Chemicals Supplier No. TRCK20080610MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56105-81-8
Molecular Formula C16 H14 O3
Molecular Weight (g/mol) 254.28
Synonym Benzeneacetic acid, 3-benzoyl-alpha-methyl-, (alphaR)-, Benzeneacetic acid, 3-benzoyl-alpha-methyl-, (R)-, (alphaR)-3-Benzoyl-alpha-methylbenzeneacetic acid, (-)-2-(3-Benzoylphenyl)propionic acid, (-)-3-Benzoyl-alpha-methylbenzeneacetic acid, (-)-Ketoprofen, (2R)-2-(3-Benzoylphenyl)propionic acid, (R)-(-)-Ketoprofen, (R)-2-(3-Benzoylphenyl)propionic acid, (R)-3-Benzoyl-alpha-methylphenylacetic acid, (R)-Ketoprofen, (R)-Ketoprophen
IUPAC Name (2R)-2-(3-benzoylphenyl)propanoic acid
SMILES C[C@@H](C(=O)O)c1cccc(c1)C(=O)c2ccccc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H14O3/c1-11(16(18)19)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-11H,1H3,(H,18,19)/t11-/m1/s1
Formula Weight 254.0943
Percent Purity >95%
Chemical Name or Material (R)-Ketoprofen
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.