missing translation for 'onlineSavingsMsg'
Learn More

(R)-Lercanidipine Hydrochloride, TRC

Product Code. 30472774
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30472774 25 mg 25mg
30470831 5 mg 5mg
30469527 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30472774 Supplier Toronto Research Chemicals Supplier No. TRCL17901025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 187731-34-6
Molecular Formula C36 H41 N3 O6 . Cl H
Molecular Weight (g/mol) 648.19
Synonym 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(3-nitrophenyl)-, 3-[2-[(3,3-diphenylpropyl)methylamino]-1,1-dimethylethyl] 5-methyl ester, hydrochloride (1:1), (4R)-, 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(3-nitrophenyl)-, 2-[(3,3-diphenylpropyl)methylamino]-1,1-dimethylethyl methyl ester, monohydrochloride, (4R)- (9CI), 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(3-nitrophenyl)-, 2-[(3,3-diphenylpropyl)methylamino]-1,1-dimethylethyl methyl ester, monohydrochloride, (R)-, (R)-Lercanidipine hydrochloride
IUPAC Name 5-O-[1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-yl] 3-O-methyl (4R)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
SMILES Cl.COC(=O)C1=C(C)NC(=C([C@@H]1c2cccc(c2)[N+](=O)[O-])C(=O)OC(C)(C)CN(C)CCC(c3ccccc3)c4ccccc4)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C36H41N3O6.ClH/c1-24-31(34(40)44-6)33(28-18-13-19-29(22-28)39(42)43)32(25(2)37-24)35(41)45-36(3,4)23-38(5)21-20-30(26-14-9-7-10-15-26)27-16-11-8-12-17-27;/h7-19,22,30,33,37H,20-21,23H2,1-6H3;1H/t33-;/m1./s1
Formula Weight 647.2762
Chemical Name or Material (R)-Lercanidipine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.