missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Linalool, 95% (sum of enantiomers)

Product Code. p-4089438 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10643415

Brand: Thermo Scientific Acros 428310250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 126-91-0 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-91-0
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00135469
InChI Key CDOSHBSSFJOMGT-JTQLQIEISA-N
Synonym --linalool, 3r-linalool, r---linalool, unii-3u21e3v8i2, 1,6-octadien-3-ol, 3,7-dimethyl-, 3r, 3r-3,7-dimethylocta-1,6-dien-3-ol, l-linalool, 3r,7-dimethylocta-1,6-dien-3-ol, r-3,7-dimethylocta-1,6-dien-3-ol, linalool +
PubChem CID 443158
ChEBI CHEBI:28
IUPAC Name (3R)-3,7-dimethylocta-1,6-dien-3-ol
SMILES CC(=CCCC(C)(C=C)O)C

Specifications

Color Colorless to Yellow
Density 0.8620g/mL
Boiling Point 196.0°C to 198.0°C
Flash Point 75°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4600 to 1.4640 (20°C, 589nm)
Quantity 25 g
Merck Index 15, 5550
Specific Gravity 0.862
Specific Rotation -15° to -21° (20°C, 589nm) (neat)
Formula Weight 154.25
Percent Purity 95% (sum of enantiomers)
Physical Form Liquid
Chemical Name or Material (R)-(-)-Linalool, 95% (sum of enantiomers)
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(-)-Linalool
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.