missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-BOC-3-Amino-3-phenylpropanoic acid, 95%, 98% ee, Thermo Scientific™

Product Code. 10170662 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10170662 500 mg Glass bottle 500mg
10055123 250 mg Glass bottle 250mg
10374212 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10170662 Supplier Thermo Scientific Acros Supplier No. 362865000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161024-80-2
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD01320859
InChI Key JTNQFJPZRTURSI-LLVKDONJSA-N
Synonym r-n-boc-3-amino-3-phenylpropanoic acid, r-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid, boc-r-3-amino-3-phenylpropionic acid, boc-beta-phe-oh, r-3-boc-amino-3-phenylpropionic acid, r-3-boc-amino-3-phenylpropanoic acid, boc-r-phenylalanine, 3r-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid, r-3-tert-butoxycarbonylamino-3-phenyl-propionic acid
PubChem CID 7009108
IUPAC Name (3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC=CC=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 123.4°C
Color White
Enantiomeric Excess 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 500 mg
Formula Weight 265.31
Percent Purity 95%,98% ee
Physical Form Powder
Chemical Name or Material (R)-N-BOC-3-Amino-3-phenylpropanoic acid, 98% ee
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.