missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-FMOC-alpha-Methylphenylalanine sesquihydrate, 98%, 98% ee, Thermo Scientific™

Product Code. 11985551 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 11985551

Brand: Thermo Scientific Acros 441071000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 152436-04-9
CAS Min % 97.5
CAS Max % 100.0
Melting Point 79°C to 104°C
Color White to Yellow
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Molecular Formula 1·5 H2O
Quantity 100 mg
Specific Rotation +28° to +32° (20°C, 589nm) (c=1 in DMF)
Synonym fmoc-alpha-methyl-d-phenylalanine, fmoc-alpha-benzyl-d-ala, r-2-9h-fluoren-9-yl methoxy carbonyl amino-2-methyl-3-phenylpropanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-2-methyl-3-phenylpropanoic acid, fmoc-alpha-methyl-d-phe, fmoc-n-a-methyl-d-phenylalanine, fmoc-, a inverted exclamation mark-benzyl-d-ala, r-2-9h-fluoren-9-yl methoxy carbonylamino-2-methyl-3-phenylpropanoic acid
InChI Key KLBKBAAOPOXFSK-RUZDIDTESA-N
SMILES CC(CC1=CC=CC=C1)(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-2-methyl-3-phenylpropanoic acid
Molecular Weight (g/mol) 428.49
PubChem CID 7010690
Formula Weight 428.49
Percent Purity 98%,98% ee
Physical Form Powder
Chemical Name or Material (R)-N-FMOC-α-Methylphenylalanine sesquihydrate, 98% ee
Show More Show Less
Product Identifier
  • (R)-N-FMOC-alpha-Methylphenylalanine sesquihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.