missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB27B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97109-50ul
This item is not returnable.
View return policy
Description
RAB27B Polyclonal antibody specifically detects RAB27B in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| RAB27B | |
| Polyclonal | |
| ELISA 1:5000-1:10000, Immunocytochemistry/ Immunofluorescence 1:300-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| C25KG, RAB27B, member RAS oncogene family, ras-related protein Rab-27B | |
| Produced in rabbits immunized with purified, recombinant Human RAB27B (Accession#: NP_004154.2; Thr2-Lys215) | |
| 50 μg | |
| Signal Transduction | |
| 5874 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction