missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB35 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49120
This item is not returnable.
View return policy
Description
RAB35 Polyclonal antibody specifically detects RAB35 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| RAB35 | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| GTP-binding protein RAY, H-ray, RAB1C, RAB35, member RAS oncogene family, Ras-related protein Rab-1C, ras-related protein rab-1c (GTP-binding protein ray), ras-related protein Rab-35, RAY | |
| This antibody was developed against a recombinant protein corresponding to amino acids: AKENVNVEEMFNCITELVLRAKKDNLAKQQQQQQNDVVKLTKNSKRKKRC | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, GPCR, Immunology, Signal Transduction | |
| 11021 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction