missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The protein encoded by this gene belongs to the RAB3 protein family, members of which are involved in regulated exocytosis of neurotransmitters and hormones. This protein forms the Rab3 GTPase-activating complex with RAB3GAP1, where it constitutes the regulatory subunit, whereas the latter functions as the catalytic subunit. This gene has the highest level of expression in the brain, consistent with it having a key role in neurodevelopment. Mutations in this gene are associated with Martsolf syndrome.
Specifications
Specifications
| Antigen | RAB3GAP2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 3A4A1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | RAB3GAP2 |
| Gene Accession No. | Q9H2M9 |
| Gene Alias | DKFZp434D245, FLJ14579, KIAA0839, p150, Rab3 GAP p150, Rab3 GAP regulatory subunit, RAB3 GAP150, RAB3GAP150, RAB3GAP2, RGAP iso |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?