Learn More
Invitrogen™ Rab4 Recombinant Superclonal™ Antibody (14HCLC)
Rabbit Recombinant Superclonal Antibody
Brand: Invitrogen™ 710543
Description
This antibody is predicted to react with bovine, canine, equine, feline, non-human primate, mouse, rabbit, rat and ovine based on sequence homology. Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.
Protein transport. Probably involved in vesicular traffic.
Specifications
| Rab4 | |
| Recombinant Superclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide | |
| P20338 | |
| Rab4a | |
| Peptide corresponding to amino acids 193-206 of human RAB4. | |
| 100 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
| Western Blot | |
| 14HCLC | |
| Unconjugated | |
| Rab4a | |
| AI848268; HRES1; HRES-1; HRES-1/RAB4; HTLV-1 related endogenous sequence; RAB4; Rab4a; RAB4A, member RAS oncogene family; ras-related protein Rab-4A; Ras-related small GTP binding protein 4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5867 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.