Learn More
Invitrogen™ RAB5 Recombinant Rabbit Monoclonal Antibody (24GB4520)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557663
 View more versions of this product
Product Code. 30275596
  • 198.00 € / 20µL
Description
RAB5 Recombinant Monoclonal Antibody for Western Blot, Flow
RAB5 are among the small GTPases Rab are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different sets of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. RAB5A is required for the fusion of plasma membranes and early endosomes. Contributes to the regulation of filopodia extension. Required for the exosomal release of SDCBP, CD63, PDCD6IP and syndecan. Regulates maturation of apoptotic cell-containing phagosomes, probably downstream of DYN2 and PIK3C3.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RAB5
Recombinant Monoclonal
0.7 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
M0RC99, P20339, Q9CQD1
RAB5A
A synthesized peptide derived from human Rab5 (aa 3-19).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB4520
Unconjugated
RAB5A
2410015H04Rik; AI663973; AU021172; nnyRab5a; Rab 5; Rab 5A; RAB5; RAB5A; RAB5A, member RAS oncogene family; Ras related protein; RAS-associated protein RAB5A; ras-related protein Rab-5A; Small GTP-binding protein rab5
Rabbit
Affinity chromatography
RUO
271457, 5868, 64633
-20°C