missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab5a Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00 € - 593.00 €
Specifications
| Antigen | Rab5a |
|---|---|
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18222571
|
Novus Biologicals
NBP2-58129 |
100 μL |
593.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18619927
|
Novus Biologicals
NBP2-58129-25ul |
25 μL |
369.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rab5a Polyclonal specifically detects Rab5a in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Rab5a | |
| Polyclonal | |
| Rabbit | |
| Endosome Markers, Signal Transduction | |
| RAB5A, member RAS oncogene family, RAB5RAS-associated protein RAB5A, ras-related protein Rab-5A | |
| RAB5A | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| 5868 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:NEPQNPGANSARGRGVDLTEPTQPTRNQCCS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title