missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB9A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 € - 326.00 €
Specifications
| Antigen | RAB9A |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18647807
|
Novus Biologicals
NBP2-99423-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18680887
|
Novus Biologicals
NBP2-99423-50ul |
50 μg |
216.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAB9A Polyclonal antibody specifically detects RAB9A in Mouse samples. It is validated for Western BlotSpecifications
| RAB9A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Signal Transduction | |
| PBS (pH 7.0) | |
| 9367 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| RAB9A, member RAS oncogene family, RAB9RAB9, member RAS oncogene family, ras-related protein Rab-9A | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human RAB9A | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title