missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AAV5 VP3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of VP1.
Brand: Abnova PAB12590.100ug
This item is not returnable.
View return policy
Specifications
| AAV5 VP3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of VP1. | |
| In PBS (0.02% sodium azide) | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Virus | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| Q9YIJ1 | |
| A synthetic peptide corresponding to amino acids 525-550 of AAV5 VP1. | |
| RUO | |
| Adeno-Associated Virus (AAV). | |
| Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction