Learn More
DPP4 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant DPP4.
Brand: Abnova PAB19140.100ug
Description
The protein encoded by this gene is identical to adenosine deaminase complexing protein-2, and to the T-cell activation antigen CD26. It is an intrinsic membrane glycoprotein and a serine exopeptidase that cleaves X-proline dipeptides from the N-terminus of polypeptides. [provided by RefSeq
Specifications
| DPP4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant DPP4. | |
| In 20mM Na-phosphate, 150mM NaCl, pH 7.5 | |
| ADABP/ADCP2/CD26/DPPIV/TP103 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 1803 | |
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western blot (1:100-1:500) Immunoprecipitation (1:10-1:40) Immunohistochemistry (1:50-1:300) The optimal working dilution should be determined by the end user. | |
| DPP4 | |
| DPP4 | |
| Recombinant protein corresponding to human DPP4 expressed in Sf 9 cells. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only