missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Goat IgG F(ab) Secondary Antibody (Pre-adsorbed), Novus Biologicals™

Product Code. 18687836 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18687836 0.5 mg 0.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18687836 Supplier Novus Biologicals Supplier No. NBP268548

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IgG F(ab) Polyclonal antibody specifically detects IgG F(ab) in Goat samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG F(ab)
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Borate buffered saline
Host Species Rabbit
Immunogen Goat IgG
Purification Method Affinity purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Goat
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.