missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETD8 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against full-length recombinant SETD8.
Brand: Abnova PAB14114.50ug
This item is not returnable.
View return policy
Specifications
| SETD8 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against full length recombinant SETD8. | |
| In PBS (0.05% sodium azide, 0.05% proclin 300) | |
| KMT5A/PR-Set7/SET07/SET8 | |
| Rabbit | |
| 50 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| SETD8 | |
| SETD8 | |
| Recombinant Flag (N-terminus) and His (C-terminus) fusion protein corresponding to full length human SETD8. | |
| RUO | |
| 387893 | |
| Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction