Learn More
STAP1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of STAP1.
Brand: Abnova PAB1880.400uL
Description
The protein encoded by this gene functions as a docking protein acting downstream of Tec tyrosine kinase in B cell antigen receptor signaling. The protein is directly phosphorylated by Tec in vitro where it participates in a postive feedback loop, increasing Tec activity. [provided by RefSeq
Specifications
| STAP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of STAP1. | |
| In PBS (0.09% sodium azide) | |
| BRDG1/STAP-1 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 26228 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:50) The optimal working dilution should be determined by the end user. | |
| STAP1 | |
| STAP1 | |
| A synthetic peptide (conjugated with KLH) corresponding to residues surrounding F56 of human STAP1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only