missing translation for 'onlineSavingsMsg'
Learn More

TAAR6 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16057687
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16057687 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16057687 Supplier Abnova Supplier No. PAB27697.50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of TAAR6.

The TAAR6 (TRAR4) gene belongs to the trace amine receptor family. Trace amines are endogenous amine compounds that are chemically similar to classic biogenic amines like dopamine, norepinephrine, serotonin, and histamine. Trace amines were thought to be 'false transmitters' that displace classic biogenic amines from their storage and act on transporters in a fashion similar to the amphetamines, but the identification of brain receptors specific to trace amines indicates that they also have effects of their own (Duan et al., 2004 [PubMed 15329799]).[supplied by OMIM

Specifications

Antigen TAAR6
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of TAAR6.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3-7 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene TAAR6
Gene Alias RP11-295F4.3/SCZD5/TA4/TRAR4
Gene Symbols TAAR6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 16 amino acid at cytoplasmic domain of human TAAR6.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 319100
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.