Learn More
Rabbit IgG VisUCyte HRP Polymer Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  R&D Systems VC003-050
 View more versions of this product
Product Code. 18768538
  • 549.00 € / 50mL
Description
IgG Polyclonal specifically detects IgG in Rabbit samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG
Polyclonal
Immunohistochemistry
Immunoglobulin G, ImmunoglobulinG
Rabbit IgG
50 mL (2 x 25 mL)
Secondary
Detects heavy ( gamma ) chains on rabbit IgG and light chains on all rabbit immunoglobulins, based on IEP. No cross-reactivity is observed with non-immunoglobulin rabbit serum proteins or serum proteins from human, mouse or rat, based on IEP. VisUCyteÖ HRP Polymer provides high sensitivity with low background in IHC.
6 months from date of receipt, 2 to 8 degreesC as supplied.
Immunohistochemistry
HRP Polymer
Supplied as a solution in PBS with preservative.
Goat
Affinity Purified
RUO
3500