missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit IgG VisUCyte HRP Polymer Antibody, R&D Systems™

Goat Polyclonal Antibody
326.00 € - 2595.00 €
Specifications
| Antigen | IgG |
|---|---|
| Dilution | Immunohistochemistry |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | HRP Polymer |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18792418
![]() |
R&D Systems
VC003-025 |
25 mL | |||||||
18768538
![]() |
R&D Systems
VC003-050 |
50 mL (2 x 25 mL) |
549.00 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18783249
![]() |
R&D Systems
VC003-125 |
125 mL |
1400.00 €
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18783868
![]() |
R&D Systems
VC003-250 |
250 mL (2 x 125 mL) |
2595.00 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgG Polyclonal specifically detects IgG in Rabbit samples. It is validated for Immunohistochemistry.Specifications
| IgG | |
| Immunohistochemistry | |
| HRP Polymer | |
| RUO | |
| Supplied as a solution in PBS with preservative. | |
| 3500 | |
| IgG | |
| Affinity Purified | |
| Detects heavy ( gamma ) chains on rabbit IgG and light chains on all rabbit immunoglobulins, based on IEP. No cross-reactivity is observed with non-immunoglobulin rabbit serum proteins or serum proteins from human, mouse or rat, based on IEP. VisUCyteÖ HRP Polymer provides high sensitivity with low background in IHC. |
| Immunohistochemistry | |
| Polyclonal | |
| Goat | |
| Rabbit | |
| Immunoglobulin G, ImmunoglobulinG | |
| Rabbit IgG | |
| Secondary | |
| 6 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title