missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit IgG VisUCyte HRP Polymer Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems VC003-250
This item is not returnable.
View return policy
Description
IgG Polyclonal specifically detects IgG in Rabbit samples. It is validated for Immunohistochemistry.
Specifications
| IgG | |
| Polyclonal | |
| Immunohistochemistry | |
| Immunoglobulin G, ImmunoglobulinG | |
| Rabbit IgG | |
| 250 mL (2 x 125 mL) | |
| Secondary | |
| Detects heavy ( gamma ) chains on rabbit IgG and light chains on all rabbit immunoglobulins, based on IEP. No cross-reactivity is observed with non-immunoglobulin rabbit serum proteins or serum proteins from human, mouse or rat, based on IEP. VisUCyteÖ HRP Polymer provides high sensitivity with low background in IHC. | |
| 6 months from date of receipt, 2 to 8 degreesC as supplied. |
| Immunohistochemistry | |
| HRP Polymer | |
| Supplied as a solution in PBS with preservative. | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3500 | |
| Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction