missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC39A7/ZIP7 Antibody [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-30747JF585
This item is not returnable.
View return policy
Description
SLC39A7/ZIP7 Polyclonal antibody specifically detects SLC39A7/ZIP7 in Human samples. It is validated for Western Blot
Specifications
| SLC39A7/ZIP7 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Janelia Fluor 585 | |
| D6S115E, D6S2244EReally interesting new gene 5 protein, H2-KE4, HKE4Solute carrier family 39 member 7, HLA class II region expressed gene KE4, KE4, Ke4 gene, mouse, human homolog of, RING5Histidine-rich membrane protein Ke4, solute carrier family 39 (zinc transporter), member 7, zinc transporter SLC39A7, ZIP7 | |
| Produced in rabbits immunized with E. coli-derived Human SLC39A7/ZIP7 fragment | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 7922 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction