missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabies Virus Nucleoprotein Antibody (02) - (CVS-11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-33165-20ul
This item is not returnable.
View return policy
Description
Rabies Virus Nucleoprotein Monoclonal antibody specifically detects Rabies Virus Nucleoprotein in Virus samples. It is validated for ELISA,Sandwich ELISA Capture
Specifications
| Rabies Virus Nucleoprotein | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rabies Virus Nucleoprotein (Uniprot#: Q8JXF6; Met1-Ser450). | |
| 20 μL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| ELISA, Sandwich ELISA Capture | |
| 02 | |
| ELISA 1:1000-1:2000, Sandwich ELISA Capture 1:250-1:2000 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction