missing translation for 'onlineSavingsMsg'
Learn More

rac 2-Hydroxy Ibuprofen, TRC

Product Code. 17980675
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17980675 10 mg 10mg
17953865 1 mg 1mg
17964175 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17980675 Supplier Toronto Research Chemicals Supplier No. TRCH94286010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51146-55-5
Molecular Formula C13 H18 O3
Molecular Weight (g/mol) 222.28
Synonym 4-(2-Hydroxy-2-methylpropyl)-α-methylbenzeneacetic acid, 2,4'-(2-Hydroxy-2-methylpropyl)phenylpropionic acid, 2-Hydroxyibuprofen, 2-[4-(2-Hydroxy-2-methylpropyl)phenyl]propanoic acid, 2-[4-(2-Hydroxy-2-methylpropyl)phenyl]propionic acid, Hydroxyibuprofen, Ibuprofen OH
IUPAC Name 2-[4-(2-hydroxy-2-methylpropyl)phenyl]propanoic acid
SMILES CC(C(=O)O)c1ccc(CC(C)(C)O)cc1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H18O3/c1-9(12(14)15)11-6-4-10(5-7-11)8-13(2,3)16/h4-7,9,16H,8H2,1-3H3,(H,14,15)
Formula Weight 222.1256
Chemical Name or Material 2-[4-(2-Hydroxy-2-methylpropyl)phenyl]propanoic Acid (2-Hydroxyibuprofen)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.