missing translation for 'onlineSavingsMsg'
Learn More

rac-4-Hydroxy Mavacamten, TRC

Product Code. 30398671
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398671 50 mg 50mg
30404364 25 mg 25mg
30397181 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398671 Supplier Toronto Research Chemicals Supplier No. TRCH29981550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-4-Hydroxy Mavacamten
Analyte or Component Names rac-4-Hydroxy Mavacamten
Shipping Condition Room Temperature
Molecular Formula C15 H19 N3 O3
InChI Formula InChI=1S/C15H19N3O3/c1-9(2)18-14(20)8-13(17-15(18)21)16-10(3)11-4-6-12(19)7-5-11/h4-10,16,19H,1-3H3,(H,17,21)
Solution Type Neat
SMILES CC(N1C(C=C(NC1=O)NC(C2=CC=C(C=C2)O)C)=O)C
IUPAC Name 6-((1-(4-hydroxyphenyl)ethyl)amino)-3-isopropylpyrimidine-2,4(1H,3H)-dione
Industry Type Pharmaceutical
Molecular Weight (g/mol) 289.33
Quantity 50 mg
Formula Weight 289.1426
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.