missing translation for 'onlineSavingsMsg'
Learn More

rac Alpha-Hydroxy Ibuprofen, TRC

Product Code. 30133920
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133920 1 mg 1mg
30003804 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133920 Supplier Toronto Research Chemicals Supplier No. TRCH9428631MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60057-62-7
Molecular Weight (g/mol) 222.28
IUPAC Name 2-hydroxy-2-[4-(2-methylpropyl)phenyl]propanoic acid
SMILES CC(C)Cc1ccc(cc1)C(C)(O)C(=O)O

Specifications

Molecular Formula C13 H18 O3
Quantity 1 mg
Synonym (2RS)-2-Hydroxy-2-[4-(2-methylpropyl)phenyl]propanoic Acid, Ibuprofen Imp. M (EP)
InChI Formula InChI=1S/C13H18O3/c1-9(2)8-10-4-6-11(7-5-10)13(3,16)12(14)15/h4-7,9,16H,8H2,1-3H3,(H,14,15)
Formula Weight 222.1256
Chemical Name or Material (2RS)-2-Hydroxy-2-[4-(2-methylpropyl)phenyl]propanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.