missing translation for 'onlineSavingsMsg'
Learn More

rac Desfluoro Citalopram Oxalate, TRC

Product Code. 30136293
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136293 5 mg 5mg
30003545 10 mg 10mg
30009889 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136293 Supplier Toronto Research Chemicals Supplier No. TRCD2899655MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1093072-86-6
Molecular Formula C20 H22 N2 O . C2 H2 O4
Molecular Weight (g/mol) 396.44
Synonym 1-(3-Dimethylaminopropyl)-1-(phenyl)-5-phthalan-carbonitrile Oxalate, 1-[3-(Dimethylamino)propyl]-1,3-dihydro-1-phenyl-5-isobenzofurancarbonitrile Ethanedioate (1:1)
IUPAC Name 1-[3-(dimethylamino)propyl]-1-phenyl-3H-2-benzofuran-5-carbonitrile;oxalic acid
SMILES CN(C)CCCC1(OCc2cc(ccc12)C#N)c3ccccc3.OC(=O)C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C20H22N2O.C2H2O4/c1-22(2)12-6-11-20(18-7-4-3-5-8-18)19-10-9-16(14-21)13-17(19)15-23-20;3-1(4)2(5)6/h3-5,7-10,13H,6,11-12,15H2,1-2H3;(H,3,4)(H,5,6)
Formula Weight 396.1685
Percent Purity >95%
Chemical Name or Material Desfluoro Citalopram Oxalate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.