missing translation for 'onlineSavingsMsg'
Learn More

rac Desmethyl Citalopram Hydrochloride, TRC

Product Code. 30362850
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362850 10 mg 10mg
30364175 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30362850 Supplier Toronto Research Chemicals Supplier No. TRCD23084610MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97743-99-2
Molecular Formula C19 H19 F N2 O . Cl H
Molecular Weight (g/mol) 346.83
Synonym (1RS)-1-(4-Fluorophenyl)-1-[3-(methylamino)propyl]-1,3-dihydroisobenzofuran-5-carbonitrile Hydrochloride, Citalopram Hydrobromide Imp. D (EP) as Hydrochloride, Desmethylcitalopram Hydrochloride, 1-(4-Fluorophenyl)-1,3-dihydro-1-[3-(methylamino)propyl]-5-isobenzofurancarbonitrile monohydrochloride, Lu 11-109C
IUPAC Name 1-(4-fluorophenyl)-1-[3-(methylamino)propyl]-3H-2-benzofuran-5-carbonitrile;hydrochloride
SMILES Cl.CNCCCC1(OCc2cc(ccc12)C#N)c3ccc(F)cc3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H19FN2O.ClH/c1-22-10-2-9-19(16-4-6-17(20)7-5-16)18-8-3-14(12-21)11-15(18)13-23-19;/h3-8,11,22H,2,9-10,13H2,1H3;1H
Formula Weight 346.1248
Percent Purity >95%
Chemical Name or Material Desmethylcitalopram Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.